• USA Home
  • Search Results

  • polyethyleneglycolmethyletheracrylate123453217139411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol) methyl ether acrylate

3 Products
Synonym:   Acryl-PEG, Methoxy PEG acrylate, Methoxy poly(ethylene glycol) monoacrylate, Poly(ethylene glycol) monomethyl ether monoacrylate, mPEG-acrylate
Image of Poly(ethylene glycol) methyl ether acrylate
CAS Number:
Linear Formula:
 H2C=CHCO2(CH2CH2O)nCH3
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Additive
  Polymer Architecture
  Reaction Suitability
  Transition temperature
  Molecular weight
  Functional group
Show More