• USA Home
  • Search Results

  • polyethyleneglycolmethyletherazide123458948561011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol) methyl ether azide

2 Products
Synonym:   α-Methoxy-ω-azido-PEG, O-(2-Azidoethyl)-O′-methylpolyethylene glycol, Methoxypolyethylene glycol azide, PEG-Azide, mPEG Azide
Image of Poly(ethylene glycol) methyl ether azide
CAS Number:
Linear Formula:
 N3CH2CH2(OCH2CH2)nOCH3
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
  Molecular weight
Show More