• USA Home
  • Search Results

  • polyethyleneglycolmethylethermethacrylate123452691572011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol) methyl ether methacrylate

5 Products
Synonym:   Methoxy PEG methacrylate, Methoxy poly(ethylene glycol) monomethacrylate, Poly(ethylene glycol) monomethyl ether monomethacrylate
Image of Poly(ethylene glycol) methyl ether methacrylate
CAS Number:
Linear Formula:
 H2C=CCH3CO2(CH2CH2O)nCH3
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
  Refractive Index
  Physical Form
  Transition temperature
  Melting Point
  Polymer Architecture
  Reaction Suitability
  Molecular weight
  Density
  Additive
Show More