• USA Home
  • Search Results

  • polyethyleneglycolphenyletheracrylate123455664105511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethylene glycol) phenyl ether acrylate

1 Products
Synonym:   Ethoxylated phenyl acrylate, Phenol ethoxylate acrylate
Image of Poly(ethylene glycol) phenyl ether acrylate
CAS Number:
Linear Formula:
 CH2CHCO2(CH2CH2O)nC6H5
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
  Refractive Index
  Additive
  Molecular weight
  Density
Show More