• USA Home
  • Search Results

  • polyethyleneglycolpolyethyleneoxidereadycalsetmp2501000000123452532268311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylene glycol/Polyethylene oxide ReadyCal Set Mp ~250 -1′000 000

1 Products
Synonym:   PEG, Poly(ethylene glycol)
Image of Polyethylene glycol/Polyethylene oxide ReadyCal Set Mp ~250 -1′000 000
CAS Number:
Linear Formula:
 H(OCH2CH2)nOH
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More