• USA Home
  • Search Results

  • polyethyleneglycolsolution123452532268311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylene glycol solution

4 Products
Synonym:   PEG solution
Image of Polyethylene glycol solution
CAS Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Application(s)
  Sterilization
  Density
  UV-Absorption Condition
  Molecular weight
  pH Value
  Special Grade
  Product Line
  Concentration
Show More