• USA Home
  • Search Results

  • polyethyleneiminesolution12345900298611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethyleneimine) solution

4 Products
Synonym:   Ethyleneimine polymer solution, PEI
Image of Poly(ethyleneimine) solution
CAS Number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Concentration
Show More