• USA Home
  • Search Results

  • polyethyleneldpe12345900288411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylene (LDPE)

1 Products
Synonym:   Polyethylene
Image of Polyethylene (LDPE)
CAS Number:
Linear Formula:
 H(CH2CH2)nH
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More