• USA Home
  • Search Results

  • polyethylenimine80ethoxylatedsolution123452665846811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylenimine, 80% ethoxylated solution

1 Products
Synonym:   PEI
Image of Polyethylenimine, 80% ethoxylated solution
CAS Number:
 MDL number:
 NACRES:
 NA.23
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Molecular weight
  Density
Show More