• USA Home
  • Search Results

  • polyethyleniminebranched12345900298611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylenimine, branched

2 Products
Synonym:   PEI
Image of Polyethylenimine, branched
CAS Number:
Linear Formula:
 H(NHCH2CH2)nNH2
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
Show More