• USA Home
  • Search Results

  • polyethyleniminelinear12345900298611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyethylenimine, linear

3 Products
Synonym:   PEI
Image of Polyethylenimine, linear
CAS Number:
Linear Formula:
 (CH2CH2NH)n
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Melting Point
  Molecular weight
Show More