• USA Home
  • Search Results

  • polyethylmethacrylate12345900342311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(ethyl methacrylate)

2 Products
Synonym:   Ethyl methacrylate homopolymer, Poly(ethyl 2-methyl-2-propeonate)
Image of Poly(ethyl methacrylate)
CAS Number:
Linear Formula:
 [CH2C(CH3)(CO2C2H5)]n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Transition temperature
  Solubility
  Density
  Physical Form
Show More