• USA Home
  • Search Results

  • polyllysinehydrobromide123452598863011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly-L-lysine hydrobromide

10 Products
Image of Poly-<SC>L</SC>-lysine hydrobromide
CAS Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Sterilization
  Solubility
  Package quantity
  Ice Shipping
  Product Line
  Color
  Physical Form
  Application(s)
Show More

Product #
Description
P2636 Poly-L-lysine hydrobromide, mol wt 30,000-70,000,
P6282 Poly-L-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture,
P9155 Poly-L-lysine hydrobromide, mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture,
P7890 Poly-L-lysine hydrobromide, mol wt 15,000-30,000 by viscosity,
P1399 Poly-L-lysine hydrobromide, mol wt 150,000-300,000,
P1274 Poly-L-lysine hydrobromide, mol wt 70,000-150,000 by viscosity,
P0879 Poly-L-lysine hydrobromide, mol wt 1,000-5,000,
P1524 Poly-L-lysine hydrobromide, mol wt ≥300,000,
P6516 Poly-L-lysine hydrobromide, mol wt 4,000-15,000 by viscosity, Green Alternative
P5899 Poly-L-lysine hydrobromide, mol wt ≥300,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture,