• USA Home
  • Search Results

  • polymethylmethacrylate12345901114711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(methyl methacrylate)

14 Products
Synonym:   PMMA, Poly(methacrylic acid methyl ester)
Image of Poly(methyl methacrylate)
CAS Number:
Linear Formula:
 [CH2C(CH3)(CO2CH3)]n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Viscosity
  Package quantity
  Manufacturer designation
  Transition temperature
  Shelf Life
  Application(s)
  Physical Form
  Anal. Standard Form
  Density
  Solubility
  Special Grade
Show More

Product #
Description
182230 Poly(methyl methacrylate), average Mw ~120,000 by GPC,
445746 Poly(methyl methacrylate), average Mw ~350,000 by GPC,
200336 Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder,
182265 Poly(methyl methacrylate),
81489 Poly(methyl methacrylate), analytical standard, for GPC, 2,000,
370037 Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical),
81501 Poly(methyl methacrylate), analytical standard, for GPC, 50,000,
03558 Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000,
81503 Poly(methyl methacrylate), analytical standard, for GPC, 100,000,
81498 Poly(methyl methacrylate), analytical standard, for GPC, 20,000,
94131 Poly(methyl methacrylate), analytical standard, for GPC, 4,000,
81497 Poly(methyl methacrylate), analytical standard, for GPC, 10,000,
81512 Poly(methyl methacrylate), analytical standard, for GPC, 8,000,
NIST1488 Poly(methyl methacrylate), NIST® SRM® 1488, 29 K narrow molecular weight distribution,