• USA Home
  • Search Results

  • polymyxinbsulfate138561140520511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polymyxin B sulfate

1 Products
Synonym:   Polymyxin B sulfate salt
Image of Polymyxin B sulfate
CAS Number:
Empirical Formula (Hill Notation):
 C55H96N16O13 · 2H2SO4
Molecular Weight:
 1385.61
 EC Number:
 MDL number:
 NACRES:
 NA.21
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More