• USA Home
  • Search Results

  • polyoxyethylene20sorbitanmonolauratesolution12345900564511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyoxyethylene (20) sorbitan monolaurate solution

1 Products
Synonym:   Polyethylene glycol sorbitan monolaurate solution, Polyoxyethylenesorbitan monolaurate
Image of Polyoxyethylene (20) sorbitan monolaurate solution
CAS Number:
 MDL number:
 NACRES
 NA.21
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  ID clarifier
Show More