• USA Home
  • Search Results

  • polypropylene12345900307011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polypropylene

4 Products
Image of Polypropylene
CAS Number:
Linear Formula:
 [CH2CH(CH3)]n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Viscosity
  Melt index
  Hardness
  Transition temperature
Show More