• USA Home
  • Search Results

  • polysodium4styrenesulfonate123452570418111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(sodium 4-styrenesulfonate)

2 Products
Synonym:   PSS, Poly(4-styrenesulfonic acid) sodium salt
Image of Poly(sodium 4-styrenesulfonate)
CAS Number:
Linear Formula:
 (C8H7NaO3S)n
 MDL number:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Melting Point
  Physical Form
  Viscosity
  Solubility
  Molecular weight
Show More