• USA Home
  • Search Results

  • polystyrenesulfonicacidsodiumsalt12345908079911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(styrenesulfonic acid sodium salt)

9 Products
Synonym:   Poly(sodium styrenesulfonate)
Image of Poly(styrenesulfonic acid sodium salt)
CAS Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Anal. Standard Form
  Physical Form
  Application(s)
  Shelf Life
  Special Grade
Show More