• USA Home
  • Search Results

  • polysulfone123452513551711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polysulfone

2 Products
Image of Polysulfone
CAS Number:
Linear Formula:
 [C6H4-4-C(CH3)2C6H4-4-OC6H4-4-SO2C6H4-4-O]n
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Viscosity
  Melt index
  Physical Form
Show More