• USA Home
  • Search Results

  • polytetrafluoroethylene12345900284011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(tetrafluoroethylene)

7 Products
Synonym:   Fluorinated polymer, Fluoropolymer, PTFE
Image of Poly(tetrafluoroethylene)
CAS Number:
Linear Formula:
 (CF2CF2)n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Transition temperature
  Density
  Particle Dimension
  Physical Form
Show More