• USA Home
  • Search Results

  • polyvinylacetate12345900320711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(vinyl acetate)

6 Products
Synonym:   Polyvinyl acetate
Image of Poly(vinyl acetate)
CAS Number:
Linear Formula:
 [CH2CH(O2CCH3)]n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Anal. Standard Form
  Manufacturer designation
  Special Grade
  Physical Form
Show More