• USA Home
  • Search Results

  • polyvinylalcohol12345900289511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(vinyl alcohol)

16 Products
Image of Poly(vinyl alcohol)
CAS Number:
Linear Formula:
 [-CH2CHOH-]n
 MDL number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Restriction
  Anal. Standard Form
  Manufacturer designation
  Application(s)
  Solubility
  Special Grade
  Viscosity
  Molecular weight
Show More

Product #
Description
341584 Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed,
360627 Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed,
P8136 Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000,
363170 Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed,
363065 Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed,
363146 Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed,
363138 Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed,
348406 Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed,
363081 Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed,
363073 Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed,
P1763 Poly(vinyl alcohol), Fully hydrolyzed,
363103 Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed,
563900 Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed,
Y0001550 Poly(vinyl alcohol), European Pharmacopoeia (EP) Reference Standard,
1.14266 Polyvinyl alcohol, protective colloid for argentometric titration,
1548065 Polyvinyl alcohol, United States Pharmacopeia (USP) Reference Standard,