• USA Home
  • Search Results

  • polyvinylalcohol268812345900289511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyvinyl alcohol 26-88

1 Products
Synonym:   PVA, PVOH, Poly(vinyl alcohol)
Image of Polyvinyl alcohol 26-88
CAS Number:
Linear Formula:
 [-CH2CHOH-]n
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
Show More