• USA Home
  • Search Results

  • polyvinylalcoholpvablendprintingfilament12345900289511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyvinyl alcohol (PVA) blend printing filament

2 Products
Synonym:   AtlasSupport, PVA blend filament
CAS Number:
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  ID clarifier
  Physical Form
  Color
Show More