• USA Home
  • Search Results

  • polyvinylchloride12345900286211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(vinyl chloride)

10 Products
Synonym:   PVC
Image of Poly(vinyl chloride)
CAS Number:
Linear Formula:
 (CH2CHCl)n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Anal. Standard Form
  Miscellaneous Specifications
  Manufacturer designation
  Special Grade
  Viscosity
  Physical Form
Show More