• USA Home
  • Search Results

  • polyvinylchloridecarboxylated123452570280111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Poly(vinyl chloride) carboxylated

4 Products
Synonym:   Poly(vinyl chloride-co-acrylic acid)
Image of Poly(vinyl chloride) carboxylated
CAS Number:
Linear Formula:
 (C3H4O2 · C2H3Cl)x
 MDL number:
 NACRES:
 NB.61
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Molecular weight
Show More