• USA Home
  • Search Results

  • polyvinylpyrrolidone12345900339811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Polyvinylpyrrolidone

13 Products
Synonym:   PVP, Polyvidone, Povidone
Image of Polyvinylpyrrolidone
CAS Number:
Linear Formula:
 (C6H9NO)n
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Molecular weight
  Restriction
  Application(s)
  Viscosity
  Special Grade
  Physical Form
Show More

Product #
Description
PVP40 Polyvinylpyrrolidone, average mol wt 40,000,
PVP10 Polyvinylpyrrolidone, average mol wt 10,000,
PVP360 Polyvinylpyrrolidone, average mol wt 360,000,
81420 Polyvinylpyrrolidone, K 30,
437190 Polyvinylpyrrolidone, average Mw ~1,300,000 by LS,
81440 Polyvinylpyrrolidone, K 90,
P0930 Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture,
234257 Polyvinylpyrrolidone, powder, average Mw ~29,000,
P2307 Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000,
856568 Polyvinylpyrrolidone, powder, average Mw ~55,000,
P5288 Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000,
90268 Polyvinylpyrrolidone, K 25, tested according to Ph Eur,
RDD033 Polyvinylpyrrolidone, free-flowing, Redi-Dri, powder, average mol wt 40,000,