• USA Home
  • Search Results

  • ponceau6r50243276677011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ponceau 6R

1 Products
Synonym:   Acid Red 44, Brilliant Crystal Scarlet, Crystal Ponceau 6R, Crystal Scarlet, Ponceau 6R
Image of Ponceau 6R
CAS Number:
Empirical Formula (Hill Notation):
 C20H12N2Na2O7S2
Molecular Weight:
 502.43
 Colour Index Number:
 16250
 E Number:
 E126
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More