• USA Home
  • Search Results

  • ponceaus76057622679511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ponceau S

2 Products
Synonym:   3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt
Image of Ponceau S
CAS Number:
Empirical Formula (Hill Notation):
 C22H12N4Na4O13S4
Molecular Weight:
 760.57
 Colour Index Number:
 27195
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Absorption
Show More