• USA Home
  • Search Results

  • potassium3methoxyphenyltrifluoroborate2140343855344711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium 3-methoxyphenyltrifluoroborate

1 Products
Image of Potassium 3-<WBR>methoxyphenyltrifluoroborate
CAS Number:
Linear Formula:
 H3COC6H4BF3 ·K
Molecular Weight:
 214.03
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More