• USA Home
  • Search Results

  • potassium4isoquinolinetrifluoroborate23506111173307311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium 4-isoquinolinetrifluoroborate

1 Products
Synonym:   Potassium isoquinoline-4-trifluoroborate
Image of Potassium 4-<WBR>isoquinolinetrifluoroborate
CAS Number:
Empirical Formula (Hill Notation):
 C9H6BF3KN
Molecular Weight:
 235.06
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More