• USA Home
  • Search Results

  • potassiumacetate981412708211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium acetate

13 Products
Image of Potassium acetate
CAS Number:
Linear Formula:
 CH3COOK
Molecular Weight:
 98.14
 Beilstein/REAXYS Number:
 3595449
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Suitability
  UV-Absorption Values
  Quality clarifier
  Purity Grade
  Melting Point
  Agency/Method
  Product Line
  Vapor pressure
  Solubility
  Physical Form
  Special Grade
  pH Value
Show More

Product #
Description
P1190 Potassium acetate, for molecular biology, ≥99.0%,
236497 Potassium acetate, ACS reagent, ≥99.0%,
791733 Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%,
W292001 Potassium acetate, 98%,
1.04820 Potassium acetate, extra pure Ph Eur,BP,JPE,E 261,
P1147 Potassium acetate, ReagentPlus®, ≥99.0%,
60035 Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT),
P5708 Potassium acetate, BioXtra, ≥99.0%,
255785 Potassium acetate, 99.98% trace metals basis,
25059 Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%,
P1222 Potassium acetate, meets USP testing specifications,
1549807 Potassium acetate, United States Pharmacopeia (USP) Reference Standard,
PX1330 Potassium Acetate, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,