• USA Home
  • Search Results

  • potassiumacetatesolution981412708211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium acetate solution

1 Products
Image of Potassium acetate solution
CAS Number:
Linear Formula:
 CH3COOK
Molecular Weight:
 98.14
 Beilstein/REAXYS Number:
 3595449
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
Show More