• USA Home
  • Search Results

  • potassiumantimonyiiitartratehydrate61383anhydrousbasis33175356111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium antimony(III) tartrate hydrate

2 Products
Synonym:   Antimony potassium tartrate
Image of Potassium antimony(III) tartrate hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C8H4K2O12Sb2 · xH2O
Molecular Weight:
 613.83 (anhydrous basis)
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More