• USA Home
  • Search Results

  • potassiumantimonyltartratetrihydrate667872830074511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium antimonyl tartrate trihydrate

3 Products
Synonym:   Antimony potassium tartrate trihydrate, Tartar emetic
Image of Potassium antimonyl tartrate trihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C8H4K2O12Sb2 · 3H2O
Molecular Weight:
 667.87
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Purity Grade
  Loss
Show More