• USA Home
  • Search Results

  • potassiumarsenatemonobasic18003778441011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium arsenate monobasic

1 Products
Synonym:   Arsenic acid potassium salt, Potassium dihydrogen arsenate
Image of Potassium arsenate monobasic
CAS Number:
Linear Formula:
 KH2AsO4
Molecular Weight:
 180.03
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More