• USA Home
  • Search Results

  • potassiumborohydride53941376251111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium borohydride

4 Products
Synonym:   Potassium tetrahydroborate, Potassium boranate
Image of Potassium borohydride
CAS Number:
Empirical Formula (Hill Notation):
 H4BK
Molecular Weight:
 53.94
 EC Index Number
 237-360-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More