• USA Home
  • Search Results

  • potassiumbromide11900775802311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium bromide

14 Products
Image of Potassium bromide
CAS Number:
Linear Formula:
 KBr
Molecular Weight:
 119.00
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 38050405
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  Greener Alternative Principles
  UV-Absorption Values
  Loss
  Quality clarifier
  Potency
  Melting Point
  Density
  Bulk Density
  Agency/Method
  Vapor pressure
  Solubility
  Product Line
  Special Grade
  pH Value
  Physical Form
Show More

Product #
Description
221864 Potassium bromide, FT-IR grade, ≥99% trace metals basis,
243418 Potassium bromide, ACS reagent, ≥99.0%, Green Alternative
1.04907 Potassium bromide, for IR spectroscopy Uvasol®,
746444 Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%, Green Alternative
1.04905 Potassium bromide, for analysis (max. 0,000001% Hg) EMSURE® ACS,Reag. Ph Eur,
P0838 Potassium bromide, BioXtra, ≥99.0%,
P9881 Potassium bromide, ReagentPlus®, ≥99.0%,
60089 Potassium bromide, BioUltra, ≥99.5% (AT),
449962 Potassium bromide, anhydrous, powder, 99.999% trace metals basis,
451010 Potassium bromide, anhydrous, powder, 99.95% trace metals basis,
793604 Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%,
1.04904 Potassium bromide, 99.999 Suprapur®,
1.04900 Potassium bromide, EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,
02110 Potassium bromide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5%,