• USA Home
  • Search Results

  • potassiumcarbonate1382158408711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium carbonate

16 Products
Synonym:   Dipotassium carbonate
Image of Potassium carbonate
CAS Number:
Linear Formula:
 K2CO3
Molecular Weight:
 138.21
 Beilstein/REAXYS Number:
 4267587
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Agency/Method
  Suitability
  UV-Absorption Values
  Purity Grade
  Potency
  Particle Dimension
  Melting Point
  Loss
  Bulk Density
  pH Value
  Product Line
  Density
  Solubility
  Quality clarifier
  Physical Form
  Assay
Show More

Product #
Description
367877 Potassium carbonate, 99.995% trace metals basis,
791776 Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%,
209619 Potassium carbonate, ACS reagent, ≥99.0%,
P5833 Potassium carbonate, BioXtra, ≥99.0%,
347825 Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh,
310263 Potassium carbonate, ReagentPlus®, 99%,
60109 Potassium carbonate, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T),
590681 Potassium carbonate, anhydrous, powder, 99.99% trace metals basis,
1.04928 Potassium carbonate, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
P1472 Potassium carbonate, meets USP testing specifications,
900502 Potassium carbonate, anhydrous, free-flowing, −325 mesh, Redi-Dri, reagent grade, ≥98%,
60108 Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T),
1.04924 Potassium carbonate, EMPROVE® ESSENTIAL Ph Eur,USP,E 501,
900501 Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%,
12611 Potassium carbonate, meets analytical specification of Ph. Helv., puriss., anhydrous, granulated, 99-101% (calc. to the dried substance),
1548167 Potassium carbonate, United States Pharmacopeia (USP) Reference Standard,