• USA Home
  • Search Results

  • potassiumchloride7455744740711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium chloride

25 Products
Image of Potassium chloride
CAS Number:
Linear Formula:
 KCl
Molecular Weight:
 74.55
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Values
  Particle Dimension
  Ice Shipping
  Suitability
  Quality clarifier
  Purity Grade
  Loss
  Application(s)
  Potency
  Bulk Density
  Solubility
  Product Line
  Special Grade
  Agency/Method
  Physical Form
  pH Value
Show More

Product #
Description
P3911 Potassium chloride, ACS reagent, 99.0-100.5%,
P9541 Potassium chloride, for molecular biology, ≥99.0%,
P9333 Potassium chloride, BioXtra, ≥99.0%,
P5405 Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%,
1.04936 Potassium chloride, for analysis EMSURE®,
P4504 Potassium chloride, ReagentPlus®, ≥99.0%,
746436 Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%,
12636 Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al,
60130 Potassium chloride, puriss. p.a., ≥99.5% (AT),
1.04938 Potassium chloride, 99.999 Suprapur®,
409316 Potassium chloride, 99.999% trace metals basis,
60128 Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT),
60126 Potassium chloride, tested according to Ph. Eur.,
1.37009 Potassium chloride, EMPROVE® EXPERT Ph Eur,BP,USP,JP,
1.04933 Potassium chloride, for analysis (<= 0.005% Br) EMSURE® ACS,ISO,Reag. Ph Eur,
793590 Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%,
204099 Potassium chloride, ≥99.99% trace metals basis,
1.04937 Potassium chloride, EMPROVE® ESSENTIAL Ph Eur,BP,E 508,
449989 Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis,
1.04935 Potassium chloride, EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,FCC,E 508,
1.04165 Potassium chloride, granular, Emprove® Expert, Ph. Eur., BP, JP, USP, New
451029 Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis,
1548190 Potassium chloride, United States Pharmacopeia (USP) Reference Standard,
PX1405 Potassium chloride, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
PHR1329 Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material,