• USA Home
  • Search Results

  • potassiumchloridesolution7455744740711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium chloride solution

9 Products
Image of Potassium chloride solution
CAS Number:
Linear Formula:
 KCl
Molecular Weight:
 74.55
 Beilstein/REAXYS Number:
 1711999
 MDL number:
 NACRES:
 ND.02
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Reaction Suitability
  UV-Absorption Condition
  Restriction
  Anal. Standard Form
  Special Grade
  Application(s)
  Quality clarifier
  Conductivity
  pH Value
  UV-Absorption Values
  Product Line
  Physical Form
  Density
Show More