• USA Home
  • Search Results

  • potassiumcyanide13c66112590968611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium cyanide-13C

1 Products
Synonym:   13C Labeled potassium cyanide, 13C labeled KCN
Image of Potassium cyanide-<SUP>13</SUP>C
CAS Number:
Linear Formula:
 K13CN
Molecular Weight:
 66.11
 Beilstein/REAXYS Number:
 3535479
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More