• USA Home
  • Search Results

  • potassiumcyanide651215150811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium cyanide

7 Products
Image of Potassium cyanide
CAS Number:
Linear Formula:
 KCN
Molecular Weight:
 65.12
 Beilstein/REAXYS Number:
 3593645
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Values
  Solubility
  Purity Grade
  Melting Point
  Density
  Bulk Density
  Agency/Method
  Potency
  Special Grade
  Physical Form
  pH Value
Show More