• USA Home
  • Search Results

  • potassiumformate841259029411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium formate

3 Products
Synonym:   Formic acid potassium salt
CAS Number:
Linear Formula:
 HCOOK
Molecular Weight:
 84.12
 Beilstein/REAXYS Number:
 3623272
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  UV-Absorption Values
Show More