• USA Home
  • Search Results

  • potassiumgluconate2342529927411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium gluconate

3 Products
Synonym:   2,3,4,5,6-Pentahydroxycaproic acid potassium salt, D-Gluconic acid potassium salt, Potassium D-gluconate
Image of Potassium gluconate
CAS Number:
Empirical Formula (Hill Notation):
 C6H11KO7
Molecular Weight:
 234.25
 Beilstein/REAXYS Number:
 3919448
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More