• USA Home
  • Search Results

  • potassiumgoldiiichloride377881368261611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium gold(III) chloride

2 Products
Synonym:   Gold potassium chloride, Potassium tetrachloroaurate(III)
Image of Potassium gold(III) chloride
CAS Number:
Linear Formula:
 KAuCl4
Molecular Weight:
 377.88
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More