• USA Home
  • Search Results

  • potassiumhexacyanoferrateiitrihydrate422391445995111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium hexacyanoferrate(II) trihydrate

5 Products
Synonym:   Potassium ferrocyanide, Yellow prussiate
Image of Potassium hexacyanoferrate(II) trihydrate
CAS Number:
Linear Formula:
 K4Fe(CN)6 · 3H2O
Molecular Weight:
 422.39
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Special Grade
Show More