• USA Home
  • Search Results

  • potassiumhydroxide5611131058311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium hydroxide

18 Products
Synonym:   Caustic potash
Image of Potassium hydroxide
CAS Number:
Linear Formula:
 KOH
Molecular Weight:
 56.11
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 38100303
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Quality clarifier
  Purity Grade
  Potency
  Agency/Method
  Product Line
  Solubility
  Special Grade
  pH Value
  Physical Form
Show More

Product #
Description
221473 Potassium hydroxide, ACS reagent, ≥85%, pellets,
P5958 Potassium hydroxide, BioXtra, ≥85% KOH basis,
484016 Potassium hydroxide, reagent grade, 90%, flakes,
306568 Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.),
1.05033 Potassium hydroxide, pellets for analysis EMSURE®,
P1767 Potassium hydroxide, ≥85% KOH basis, pellets, white,
06103 Potassium hydroxide, technical, ≥85%, powder,
757551 Potassium hydroxide, anhydrous, ≥99.95% trace metals basis,
1.05012 Potassium hydroxide, pellets EMPLURA®,
30603 Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%,
06005 Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets,
1.05029 Potassium hydroxide, pellets for analysis (max. 0.05% Na) EMSURE® ACS,Reag. Ph Eur,
909122 Potassium hydroxide, free-flowing, Redi-Dri, reagent grade, 90%, flakes,
60377 Potassium hydroxide, tested according to Ph. Eur.,
1.05032 Potassium hydroxide, pellets EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,FCC,E 525,
60370 Potassium hydroxide, puriss. p.a., ≥86% (T), pellets,
PX1480 Potassium hydroxide, pellets, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
911062 Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets,