• USA Home
  • Search Results

  • potassiumiodate21400775805611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Potassium iodate

7 Products
Image of Potassium iodate
CAS Number:
Empirical Formula (Hill Notation):
 IKO3
Molecular Weight:
 214.00
 EC Index Number
 231-831-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Physical Form
  Solubility
  Product Line
  Loss
  pH Value
  Special Grade
Show More